Learn More
Scientific Industries SI™ Replacement Platform with Rubber Cover
Product Code. 15557315
Click to view available options
For Use With (Equipment):
Vortex-Genie family of mixers
Unit Size:
Each
Product Code. 15557315
Brand: Scientific Industries SI™ 83041423
Only null left
Add to basket
Edge
Add to basket
Fits Vortex-Genie 1, Vortex-Genie Pulse and the Vortex-Genie 2 family of mixers. Scientific Industries SI™ Replacement Platform without Rubber Cover is for use with vortexing in beakers, flasks and multiple tubes.
  • For use with Vortex-Genie family of products
  • Cover not included
Specifications
Material Molded rubber
For Use With (Equipment) Vortex-Genie family of mixers