Learn More
Thermo Scientific™ Plate Count Agar (Bottled)
Description
Enumerate viable organisms in milk, dairy products, water, food and other specimens of sanitary importance using non-selective Thermo Scientific™ Oxoid™ Plate Count Agar ready-prepared bottled medium. Plate Count Agar is equivalent to the medium recommended by the APHA1, PHLS 2 and ISO3,4 for the plate count of microorganisms in food.
Remove the need for autoclaving and dispensing with convenient, pre-filled and ready-to-use bottles of Plate Count Agar.
- Convenient: pre-filled bottles are ready for immediate use.
- Simple: easy-to-use with minimum training.
- Long shelf life: upto 12 months.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
Specifications
Specifications
| Color | Straw, Clear |
| Content And Storage | 2 to 25°C |
| Description | Plate Count Agar(Bottled) |
| Format | Sirop Bottle |
| Manufacturing Origin | United Kingdom |
| Product Type | Agar |
| Fill Weight | 500.0g + 4.0g |
| Packaging Type | 10 bottles with screw cap closures in a box |
| Quantity | 10/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.