missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH3GL3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA568330
This item is not returnable.
View return policy
Description
Predicted to react with Human and Rat samples.
SH3GL3 Polyclonal antibody specifically detects SH3GL3 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| SH3GL3 | |
| Polyclonal | |
| Unconjugated | |
| Sh3gl3 | |
| CNSA3; EEN-B2; Endophilin 3; endophilin A3; endophilin III; endophilin-3; Endophilin-A3; HsT19371; SH3 domain containing GRB2 like 3; SH3 domain containing GRB2 like 3, endophilin A3; SH3 domain containing GRB2 like endophilin A3; SH3 domain protein 2 C1; SH3 domain protein 2 C2; SH3 domain protein 2C; SH3 domain-containing GRB2-like protein 3; Sh3d2c; Sh3d2c1; Sh3d2c2; SH3-domain GRB2-like 3; Sh3gl3; SH3p13 | |
| Rabbit | |
| affinity chromatography | |
| RUO | |
| 20408, 6457, 81921 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O35180, Q62421, Q99963 | |
| Sh3gl3 | |
| Synthesized peptide derived from human SH3GL3. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction