missing translation for 'onlineSavingsMsg'
Learn More

Platinum (0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex, soln. in vinyl terminated polydimethylsiloxane

Product Code. 11304258
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11304258

Brand: Thermo Scientific Alfa Aesar 041508.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 68478-92-2 | C8 H18 O Pt Si2 | 474.68 g/mol

Catalyst for Si-H addition to olefins silicone vinyl addition room temperature cure catalyst.Platinum (0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex, soln. in vinyl terminated polydimethylsiloxane is used as a catalyst in hydrosilylation reaction for the preparation of trans-beta-silanes and in reductive amination reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Catalyst for Si-H addition to olefins silicone vinyl addition room temperature cure catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68478-92-2
Molecular Formula C8 H18 O Pt Si2
Molecular Weight (g/mol) 474.68
MDL Number MFCD00151662
InChI Key RCNRJBWHLARWRP-UHFFFAOYSA-N
Synonym platinum 0-1,3-divinyl-1,1,3,3-tetramethyldisiloxane, 1,1,3,3-tetramethyl-1,3-divinyldisiloxane, platinum salt, platinum 0-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex solution, ksc628a5r, platinum 1,3-divinyl-1,1,3,3-tetramethyldisiloxane, dimethyl vinyl silyl oxy-dimethyl-vinyl-silane; platinum, ethenyl ethenyldimethylsilyl oxy dimethylsilane platinum, platinum 0-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex, platinum,1,3-diethenyl-1,1,3,3-tetramethyldisiloxane complexes
PubChem CID 10959889
SMILES [Pt].C[Si](C)(O[Si](C)(C)C=C)C=C

Specifications

Color Orange
Density 0.984 g/mL
Flash Point >110°C (230°F)
Quantity 25 g
Sensitivity Moisture sensitive
Solubility Information Miscible with toluene,xylene,isopropyl alcohol,double-vinyl silicone oil and vinyl terminated poly(dimethylsiloxane). Immiscible with water.
IUPAC Name ethenyl-[ethenyl(dimethyl)silyl]oxy-dimethylsilane;platinum
Formula Weight 474.68
Physical Form Liquid
Chemical Name or Material Platinum (0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex, Soln. in vinyl terminated polydimethylsiloxane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.