Learn More
Platinum-Cobalt (Hazen) 500-Color Standard, Reagecon
Used to calibrate, control, qualify or validate color measurement instruments
426.00 €
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12077276
|
Reagecon
HAZ500 |
1 L | HDPE Bottle GL45 |
426.00 €
1 L |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Platinum-Cobalt (Hazen) Color Reference Standard, Platinum-Cobalt (Hazen) Color Reference Material
- This standard can be described as Platinum-Cobalt, Pt-Co, Hazen or APHA color.
- All descriptions are interchangeable.
- This product is used extensively in the water industry and several other industries.
- The standard covers the requirements of ASTM Method D 1209.
- This product is prepared gravimetrically on a weight/weight basis.
- Both solute and solvent are weighed on a balance calibrated by Reagecon engineers using OIML traceable weights.
- Reagecon holds ISO 17025 accreditation for calibration of laboratory balances (INAB Ref: 265C).
- The resulting Balance Certificate of Calibration is issued in accordance with the requirements of ISO/IEC 17025.
- The concentration of each standard is verified using a high-performance top of the range calibrated spectrophotometer.
- The calibration of the spectrophotometer is controlled using high purity ISO Guide 34 accredited spectrophotometric standards.