missing translation for 'onlineSavingsMsg'
Learn More

Platinum, plasma standard solution, Specpure™ Pt 10,000μg/mL

Product Code. 11440430
Change view
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Unit Size:
100mL
500mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440430 500 mL 500mL
11392318 50 mL 50mL
11302328 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11440430 Supplier Thermo Scientific Alfa Aesar Supplier No. 014397.AP
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7440-6-6 | Pt in 20% HCl

Platinum, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6-6-7440
Molecular Formula Pt in 20% HCl
MDL Number MFCD00011179

Specifications

Odor Characteristic
Physical Form Liquid
Solubility Miscible with water
Type Analytical Standard
Quantity 500 mL
Grade Specpure
Chemical Name or Material Platinum

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.