missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLC-gamma 2 Antibody (1E10C11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
217.00 € - 578.00 €
Specifications
| Antigen | PLC-gamma 2 |
|---|---|
| Clone | 1E10C11 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18207280
|
Novus Biologicals
NBP2-52535 |
0.1 mg |
578.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18664578
|
Novus Biologicals
NBP2-52535-0.025mg |
0.025 mg |
217.00 €
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PLC-gamma 2 Monoclonal specifically detects PLC-gamma 2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| PLC-gamma 2 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| Unconjugated | |
| Mouse | |
| Lipid and Metabolism, Phospho Specific | |
| PBS with 0.05% Sodium Azide | |
| 5336 | |
| Purified recombinant fragment of human PLCG2 (AA: 826-985) expressed in E. Coli. | |
| Primary | |
| 147.9 kDa |
| 1E10C11 | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 3.1.4.11, Phosphoinositide phospholipase C-gamma-2, phospholipase C gamma 2, phospholipase C, gamma 2 (phosphatidylinositol-specific), phospholipase C, gamma 2 (phosphatidylyinositol-specific), Phospholipase C-gamma-2,1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase gamma-2, Phospholipase C-IV, PLCG2, PLC-gamma-2, PLC-IV | |
| PLCG2 | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title