missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLC-gamma 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-86030-25ul
This item is not returnable.
View return policy
Description
PLC-gamma 2 Polyclonal antibody specifically detects PLC-gamma 2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| PLC-gamma 2 | |
| Polyclonal | |
| Western Blot 0.04 - 0.4 ug/mL, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50-1:200 | |
| EC 3.1.4.11, Phosphoinositide phospholipase C-gamma-2, phospholipase C gamma 2, phospholipase C, gamma 2 (phosphatidylinositol-specific), phospholipase C, gamma 2 (phosphatidylyinositol-specific), Phospholipase C-gamma-2,1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase gamma-2, Phospholipase C-IV, PLCG2, PLC-gamma-2, PLC-IV | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: FGDLLLTKPTEASADQLPSPSQLREKIIIKHKKLGPRGDVDVNMEDKKDEHKQQGELYMWDSIDQKWTRHYCAIADAKLSFSDDIEQTMEEEVPQDIPPTELHFGEKWFHKK | |
| 25 μL | |
| Lipid and Metabolism | |
| 5336 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction