missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLD6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-13771
This item is not returnable.
View return policy
Description
PLD6 Polyclonal specifically detects PLD6 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| PLD6 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Choline phosphatase 6, EC 3.1.4.-, Mitochondrial phospholipase, MitoPLD, Phosphatidylcholine-hydrolyzing phospholipase D6, phospholipase D family, member 6, Phospholipase D6, PLD 6, Protein zucchini homolog | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| PLD6 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: QDPGYMHHKFAIVDKRVLITGSLNWTTQAIQNNRENVLITEDDEYVRLFLEEFERIWEQFNPTKYTFFPPKKSHGSCAPPVSRAGGRLLSWHRTCGTSSESQT | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 201164 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction