missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: QQVHTIQEFV HHDENHSSPS AENMNEASSL LSATCNTFIT TLEECVKIAN AKFKPEMFQL HHPDPLVSPV SPSPVQMMKR SVSHP Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 95%.
PLEKHA3 (Pleckstrin Homology Domain Containing A3) is a Protein Coding gene. Among its related pathways are Metabolism and Cholesterol and Sphingolipids transport / Transport from Golgi and ER to the apical membrane (normal and CF). Gene Ontology (GO) annotations related to this gene include phospholipid binding and 1-phosphatidylinositol binding. An important paralog of this gene is PLEKHA8.
Specifications
Specifications
| Antigen | PLEKHA3 |
| Applications | Immunocytochemistry, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Plekha3 |
| Gene Accession No. | Q9HB20 |
| Gene Alias | FAPP1; FAPP-1; four-phosphate-adaptor protein 1; PH domain-containing family A member 3; Phosphatidylinositol-four-phosphate adapter protein 1; phosphoinositol 4-phosphate adapter protein 1; pleckstrin homology domain containing A3; pleckstrin homology domain containing, family A (phosphoinositide binding specific) member 3; pleckstrin homology domain-containing family A member 3; pleckstrin homology domain-containing, family A (phosphoinositide binding specific) member 3; Plekha3 |
| Gene Symbols | Plekha3 |
| Show More |
Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?