Learn More
PLIN5 Monoclonal Antibody (OTI3G4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA811701
Encompass_Preferred
 View more versions of this product
Product Code. 17505884
  • 540.87 € / 100µL
Description
Lipid droplet-associated protein that maintains the balance between lipogenesis and lipolysis and also regulates fatty acid oxidation in oxidative tissues. Recruits mitochondria to the surface of lipid droplets and is involved in lipid droplet homeostasis by regulating both the storage of fatty acids in the form of triglycerides and the release of fatty acids for mitochondrial fatty acid oxidation. In lipid droplet triacylglycerol hydrolysis, plays a role as a scaffolding protein for three major key lipolytic players: ABHD5, PNPLA2 and LIPE. Reduces the triacylglycerol hydrolase activity of PNPLA2 by recruiting and sequestering PNPLA2 to lipid droplets. Phosphorylation by PKA enables lipolysis probably by promoting release of ABHD5 from the perilipin scaffold and by facilitating interaction of ABHD5 with PNPLA2. Also increases lipolysis through interaction with LIPE and upon PKA-mediated phosphorylation of LIPE.
Specifications
Specifications
PLIN5
Monoclonal
Unconjugated
PLIN5
LSDA5, LSDP5, MLDP, OXPAT
Mouse
Affinity Chromatography
RUO
440503
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry (Paraffin), Western Blot
OTI3G4
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q00G26
PLIN5
Human recombit protein fragment corresponding to amino acids 1-463 of human PLIN5 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2a