missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLK1 Antibody (AZ24) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2792
This item is not returnable.
View return policy
Description
PLK1 Monoclonal antibody specifically detects PLK1 in Human, Mouse, Xenopus samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
| PLK1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500 | |
| cell cycle regulated protein kinase, EC 2.7.11, EC 2.7.11.21, PLK, PLK-1, polo (Drosophia)-like kinase, polo like kinase, polo-like kinase 1polo-like kinase (Drosophila), Serine/threonine-protein kinase 13, serine/threonine-protein kinase PLK1, STPK13 | |
| Baculo-expressed PLK1 from Xenopus laevis | |
| 0.1 mg | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Repair, Mitotic Regulators, Stem Cell Markers | |
| 5347 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, ELISA, Immunoprecipitation | |
| AZ24 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Xenopus species | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction