missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLK1 Antibody (CL8360) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21166JF525
This item is not returnable.
View return policy
Description
PLK1 Monoclonal antibody specifically detects PLK1 in Human samples. It is validated for Western Blot
Specifications
| PLK1 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| cell cycle regulated protein kinase, EC 2.7.11, EC 2.7.11.21, PLK, PLK-1, polo (Drosophia)-like kinase, polo like kinase, polo-like kinase 1polo-like kinase (Drosophila), Serine/threonine-protein kinase 13, serine/threonine-protein kinase PLK1, STPK13 | |
| This antibody was developed using a synthetic peptide derived from P53350, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Mitotic Regulators, Phospho Specific, Stem Cell Markers | |
| 5347 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Western Blot | |
| CL8360 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction