missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLK1 Antibody (CL8360), Novus Biologicals™
Mouse Monoclonal Antibody
329.00 € - 516.00 €
Specifications
| Antigen | PLK1 |
|---|---|
| Clone | CL8360 |
| Dilution | Western Blot 1 ug/ml |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18636284
|
Novus Biologicals
NBP3-21166-100ul |
100 μg |
516.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18604614
|
Novus Biologicals
NBP3-21166-25ul |
25 μg |
329.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PLK1 Monoclonal antibody specifically detects PLK1 in Human samples. It is validated for Western BlotSpecifications
| PLK1 | |
| Western Blot 1 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| cell cycle regulated protein kinase, EC 2.7.11, EC 2.7.11.21, PLK, PLK-1, polo (Drosophia)-like kinase, polo like kinase, polo-like kinase 1polo-like kinase (Drosophila), Serine/threonine-protein kinase 13, serine/threonine-protein kinase PLK1, STPK13 | |
| This antibody was developed against a Synthetic Peptide of P53350. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| CL8360 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Mitotic Regulators, Phospho Specific, Stem Cell Markers | |
| PBS, pH 7.2, 40% glycerol | |
| 5347 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title