Learn More
Invitrogen™ PLSCR3 Recombinant Rabbit Monoclonal Antibody (24GB1345)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557582
 View more versions of this product
Product Code. 30275235
  • 564.00 € / 100µL
Description
PLSCR3 Recombinant Monoclonal Antibody for Western Blot, Flow
May mediate accelerated ATP-independent bidirectional transbilayer migration of phospholipids upon binding calcium ions that results in a loss of phospholipid asymmetry in the plasma membrane. May play a central role in the initiation of fibrin clot formation, in the activation of mast cells and in the recognition of apoptotic and injured cells by the reticuloendothelial system. Seems to play a role in apoptosis, through translocation of cardiolipin from the inner to the outer mitochondrial membrane which promotes BID recruitment and enhances tBid-induced mitochondrial damages. [UniProt].
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PLSCR3
Recombinant Monoclonal
Unconjugated
Plscr3
2210403O21Rik; 2610037N06Rik; Ca 2+ dependent phospholipid scramblase 3; ca(2+)-dependent phospholipid scramblase 3; ESTM3; LOW QUALITY PROTEIN: phospholipid scramblase 3; LOW QUALITY PROTEIN: phospholipid scramblase 3; phospholipid scramblase 3; phospholipid scramblase 3; PL scramblase 3; Pls3; Plscr3; Unknown (protein for MGC:138060); X83310
Rabbit
Affinity chromatography
RUO
360549, 57048, 70310
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB1345
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q6QBQ4, Q9JIZ9, Q9NRY6
Plscr3
A synthesized peptide derived from human PLSCR3 (aa 1-45).
100 μL
Primary
Human, Mouse, Rat