missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Memmert™ Plug-In Tube Extension for SF260, Straight
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
SF260 Sterilizer
Unit Size:
Each
Description
- Accessory
- Plug-in tube extension (outer diameter 60.3 mm, inner 57 mm)
- Straight
Specifications
Specifications
| Diameter (Metric) Outside | 60.3 mm |
| For Use With (Equipment) | SF260 Sterilizer |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction