Learn More
Memmert™ Plug-In Tube Extension for SN260, Straight
Product Code. 17508535 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
SN260 Sterilizer
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
17508535 SN260 Sterilizer Each
1 options
Product Code. 17508535 Supplier Memmert™ Supplier No. SN260B29718
Add to basket
Add to basket
Plug-in tube extension designed for exhaust air ducting and connection by hose if necessary.
  • Accessory
  • Plug-in tube extension (outer diameter 60.3 mm, inner 57 mm)
  • Straight
Specifications
Diameter (Metric) Outside 60.3 mm
For Use With (Equipment) SN260 Sterilizer