missing translation for 'onlineSavingsMsg'
Learn More

Plumbagin, 99+%

Product Code. 10235810 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 10235810

Brand: Thermo Scientific Acros 184040010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 481-42-5 | C11H8O3 | 188.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 481-42-5
Melting Point 75°C to 78°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Molecular Formula C11H8O3
MDL Number MFCD00001682
Quantity 1 g
Merck Index 15, 7655
Synonym plumbagin, plumbagine, 5-hydroxy-2-methyl-1,4-naphthoquinone, plumbaein, plumbagone, 2-methyljuglone, 5-hydroxy-2-methyl-1,4-naphthalenedione, 2-methyl-5-hydroxy-1,4-naphthoquinone, 5-hydroxy-2-methyl-1,4 naphthoquinone, 1,4-naphthalenedione, 5-hydroxy-2-methyl
Solubility Information Solubility in water: slightly soluble in hot water. Other solubilities: soluble in alcohol,acetone,chloroform,benzene,,acetic acid
InChI Key VCMMXZQDRFWYSE-UHFFFAOYSA-N
SMILES CC1=CC(=O)C2=C(C1=O)C=CC=C2O
IUPAC Name 5-hydroxy-2-methylnaphthalene-1,4-dione
Molecular Weight (g/mol) 188.18
PubChem CID 10205
ChEBI CHEBI:8273
Formula Weight 188.18
Percent Purity 99+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Plumbagin, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • Plumbagin
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.