missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PMCA4 ATPase Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA583500

Product Code. 16350455

  • 524.00 € / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: THPEFAIEEE LPRTPLLDEE EEENPDKASK FGTRVLLLDG EVTPYANTNN NAVDCNQVQL PQSDSSLQSL ETSV Highest antigen sequence indentity to the following orthologs: Rat - 46%, Mouse - 53%.

The calcium pump of the plasma membrane, termed PMCA ATPase, pumps calcium from the cytosol to the extracellular space. This membrane-bound enzyme is related to a number of other ATPases including the SERCA ATPase and the sodium/K+ pump. There are four different genes encoding PMCA ATPase and studies have revealed 20 isoforms of the pump generated by alternate splicing of the primary gene products. mRNA distribution studies show that gene products 1 and 4 are transcribed in most tissues, however, products 2 and 3 are more tissue specific. Transcription of the splicing variants has also been found to be tissue specific. In the pancreas, where insulin secretion is calcium dependent, the beta cells only express the 4b isoform, however the alpha and gamma cells express both 4a and 4b isoforms. Studies have also shown that different splice variants have different affinities for calcium and calmodulin (Cat. # MA3-917 and MA3-918).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PMCA4 ATPase
Polyclonal
Unconjugated
Atp2b4
ATP2B2; ATP2B4; ATPase plasma membrane Ca2+ transporting 4; ATPase, Ca++ transporting, plasma membrane 4; ATPase, Ca++ transporting, plasma membrane 4 ATP synthase, H+ transporting, mitochondrial F1; ATPase, Ca++ transporting, plasma membrane 4 tv2; matrix-remodeling-associated protein 1; MXRA1; plasma membrane Ca2+-ATPases 4; plasma membrane calcium ATPase; plasma membrane calcium ATPase 4; plasma membrane calcium ATPase 4 variant b; Plasma membrane calcium ATPase isoform 4; plasma membrane calcium pump; Plasma membrane calcium pump isoform 4; plasma membrane calcium-pumping ATP-ase; plasma membrane calcium-transporting ATPase 4; PMCA4; PMCA4b; PMCA4x; sarcolemmal calcium pump
Rabbit
Antigen affinity chromatography
RUO
493
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry (Paraffin)
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P23634
Atp2b4
Recombinant protein corresponding to Human ATP2B4. Recombinant protein control fragment (Product #RP-95778).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.