Learn More
Invitrogen™ PMF1 Recombinant Rabbit Monoclonal Antibody (24GB985)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557612
 View more versions of this product
Product Code. 30275565
  • 564.00 € / 100µL
Description
PMF1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
PMF1 is part of the MIS12 complex which is required for normal chromosome alignment and segregation and kinetochore formation during mitosis. It may act as a cotranscription partner of NFE2L2 involved in regulation of polyamine-induced transcription of SSAT.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PMF1
Recombinant Monoclonal
Unconjugated
PMF1
2600009M07Rik; AL033286; AW060657; Est1p-like protein B (EST1B); Pmf1; PMF-1; polyamine modulated factor 1; polyamine-modulated factor 1; RP11-54H19.4; Unknown (protein for MGC:134238)
Rabbit
Affinity chromatography
RUO
11243, 67037, 681050
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB985
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q6P1K2, Q9CPV5
PMF1
A synthesized peptide derived from human PMF1 (aa 1-205).
100 μL
Primary
Human, Mouse, Rat