missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PMFBP1 Monoclonal Antibody (4G9)

Description
Peptide Sequence: FHTEELQTSY YSLRQYQSIL EKQTSDLVLL HHHCKLKEDE VILYEEEMGN HNENTGEKLH LAQEQLALAG DKIASLERSL NLYRDKYQSS LSNIELLEC.
May play a role in sperm morphology especially the sperm tail and consequently affect fertility. May also be involved in the general organization of cellular cytoskeleton.
Specifications
Specifications
| Antigen | PMFBP1 |
| Applications | ELISA, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 4G9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | PMFBP1 |
| Gene Accession No. | Q8TBY8 |
| Gene Alias | 1700016D22Rik; F77; LOW QUALITY PROTEIN: polyamine-modulated factor 1-binding protein 1; ODF3; outer dense fiber ODF3; outer dense fiber protein 3; PMF-1 binding protein; PMF-1-binding protein; PMFBP1; polyamine modulated factor 1 binding protein 1; polyamine-modulated factor 1-binding protein 1; sperm tail associated protein; sperm tail-associated protein; Stap |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?