missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PML Protein Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98823-100ul
This item is not returnable.
View return policy
Description
PML Protein Polyclonal antibody specifically detects PML Protein in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| PML Protein | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:300-1:10000, Immunoprecipitation 4-8 μL/mg of lysate | |
| MYLPP8675, promyelocytic leukemia, Promyelocytic leukemia protein, RING finger protein 71, RNF71probable transcription factor PML, TRIM19promyelocytic leukemia, inducer of, tripartite motif protein TRIM19, Tripartite motif-containing protein 19 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human PML Protein. | |
| 100 μg | |
| Cancer, Zinc Finger | |
| 5371 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction