missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PMP22 Antibody (SR3611), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27787-100ul
This item is not returnable.
View return policy
Description
PMP22 Monoclonal antibody specifically detects PMP22 in Human,Rat samples. It is validated for Western Blot
Specifications
| PMP22 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot | |
| SR3611 | |
| Western Blot 1:500-1:2000 | |
| CMT1A, CMT1E, GAS3, GAS-3DSS, growth arrest-specific 3, Growth arrest-specific protein 3, HMSNIA, HNPP, MGC20769, peripheral myelin protein 22, PMP-22, Sp110 | |
| A synthesized peptide derived from human PMP22 (Uniprot #: Q01453) | |
| 100 μL | |
| Cellular Markers, Glia Markers, Membrane Trafficking and Chaperones, Neuroscience, Oligodendrocyte Cell Markers | |
| 5376 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction