missing translation for 'onlineSavingsMsg'
Learn More

PMSF, MedChemExpress

Product Code. 16428346
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16428346 100 mg 100mg
16402166 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16428346 Supplier Medchem Express Supplier No. HYB0496
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress PMSF is an irreversible serine/cysteine protease inhibitor commonly used in the preparation of cell lysates.

PMSF is an irreversible serine/cysteine protease inhibitor commonly used in the preparation of cell lysates.

Specifications

Chemical Name or Material PMSF
Physical Form Solid
CAS 329-98-6
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C7H7FO2S
Synonym Phenylmethylsulfonyl fluoride Benzylsulfonyl fluoride
Solubility Information DMSO : 250 mg/mL (1435.21 mM; Need ultrasonic) ∣Isopropanol : 60 mg/mL (344.45 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=S(CC1=CC=CC=C1)(F)=O
Molecular Weight (g/mol) 174.19
Formula Weight 174.19
Percent Purity 98.02%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.