missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PNMA1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15594302
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15594302 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15594302 Supplier Invitrogen™ Supplier No. PA540648

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: KSNNPAITTA ECLKALEQVF GSVESSRDAQ IKFLNTYQNP GEKLSAYVIR Sequence homology: Cow: 79%; Dog: 86%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 86%; Rabbit: 86%; Rat: 86%.

PNMA1 encodes a protein that is highly restricted to the brain and testis. Anti-PNMA1 reacts mainly with subnuclear elements (including the nucleoli) and to a lesser degree the cytoplasm.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PNMA1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Pnma1
Gene Accession No. Q8ND90
Gene Alias 37 kDa neuronal protein; 5730402C15Rik; MA1; Neuron- and testis-specific protein 1; paraneoplastic antigen MA1; Paraneoplastic antigen Ma1 homolog; paraneoplastic Ma antigen 1; paraneoplastic neuronal antigen MA1; paraneoplastic onconeuronal protein MA1; PNMA family member 1; PNMA1
Gene Symbols Pnma1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human PNMA1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9240
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.