missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ POC5 Recombinant Protein Antigen

Product Code. 18203899 Shop All Bio Techne Products
Click to view available options
Formulation:
Amino Acid Sequence: KWEEYEELLHYAIVTPNIEPCASQSSHPKGELVPDVRISTIHDILHSQGNNSEVRETAIEVGKGCDFHISSHSKTDESSPVLSPRKPSHPVMD
Amino Acid Sequence: YVPRVVTSAQQKAGRTITARITGRCDFASKNRISSSLAIMGVSPPMSSVVVEKHHPVTVQTIPQATAAKYPRTIHPESSTSASRSLGTRSAHTQSLTSVH
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 18203899

Brand: Novus Biologicals™ NBP276534PEP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Antibody Competition

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human POC5

The POC5 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 134359
Species Human
Purity >80% by SDS-PAGE and Coomassie blue staining
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Formulation Amino Acid Sequence: KWEEYEELLHYAIVTPNIEPCASQSSHPKGELVPDVRISTIHDILHSQGNNSEVRETAIEVGKGCDFHISSHSKTDESSPVLSPRKPSHPVMD
For Use With (Application) Antibody Competition
Gene Symbol POC5
Label Type Unconjugated
Molecular Weight (g/mol) 28 kDa
Quantity 100 μL
Regulatory Status RUO
Source E. coli
Specific Reactivity POC5
Storage Buffer PBS and 1M Urea, pH 7.4.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.