Learn More
Podocalyxin Like Antibody (CL0285) [mFluor Violet 500 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52904MFV500
Product Code. 30305784
  • 759.00 € / 0.10mL
Description
Podocalyxin Like Monoclonal antibody specifically detects Podocalyxin Like in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Podocalyxin Like
Monoclonal
mFluor Violet 500 SE
Gp200MGC138240, PCLP1, PCLP-1, PCLPGCTM-2 antigen, PCpodocalyxin, podocalyxin-like, Podocalyxin-like protein 1
This antibody was developed using a recombinant protein derived from O00592, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Cell Biology, Cellular Markers, Hematopoietic Stem Cell Markers, Stem Cell Markers
5420
Store at 4⪚C in the dark.
IgG2b
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL0285
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified