Learn More
Invitrogen™ POLB Recombinant Rabbit Monoclonal Antibody (24GB1475)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557383
 View more versions of this product
Product Code. 30276544
  • 564.00 € / 100µL
Description
POLB Recombinant Monoclonal Antibody for Western Blot
DNA polymerase beta comprises an amino-terminal 8- kDa domain and a carboxy-terminal 31- kDa domain. The N-terminal ssDNA binding domain has a deoxyribose phosphodiesterase activity while the C-terminal domain has a nucleotidyltransferase activity. Mammalian DNA polymerase beta, a DNA repair polymerase, is constitutively expressed in cultured cells, but treatment of cells with the DNA-alkylating agents such as N-methyl-N'-nitro-N-nitrosoguanidine (MNNG) or methyl methanesulfonate up-regulates beta-pol level. DNA polymerase beta fills single nucleotide gaps in DNA produced by the base excision repair pathway of mammalian cells.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
POLB
Recombinant Monoclonal
Unconjugated
POLB
A430088C08Rik; DNA pol beta; DNA polymerase beta; DNA polymerase beta subunit; DNA-directed DNA polymerase beta; hypothetical protein LOC614688; LOW QUALITY PROTEIN: DNA polymerase beta; MGC125976; POL B; Pol beta; POLB; polb.S; polymerase (DNA directed), beta; polymerase (DNA directed), beta S homeolog; polymerase (DNA) beta; polymerase B; polymerase-beta; wu:fa97a05; XELAEV_18020165mg; zgc:109924
Rabbit
Affinity chromatography
RUO
18970, 29240, 5423
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB1475
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P06746, P06766, Q8K409
POLB
A synthesized peptide derived from human DNA polymerase beta (aa 300-335).
100 μL
Primary
Human, Mouse, Rat