missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POLD1 Recombinant Rabbit Monoclonal Antibody (24GB4155)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557178
This item is not returnable.
View return policy
Description
POLD1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes the 125-kDa catalytic subunit of DNA polymerase delta. DNA polymerase delta possesses both polymerase and 3' to 5' exonuclease activity and plays a critical role in DNA replication and repair. Alternatively spliced transcript variants have been observed for this gene, and a pseudogene of this gene is located on the long arm of chromosome 6.
Specifications
| POLD1 | |
| Recombinant Monoclonal | |
| 1.5 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O54747, P28340, P52431 | |
| POLD1 | |
| A synthesized peptide derived from human POLD1 (aa 1000-1065). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB4155 | |
| Unconjugated | |
| POLD1 | |
| 125kDa; CDC2; CDC2 homolog; CRCS10; DAN polymerase delta 1, catalytic subunit; DNA polymerase delta 1, catalytic domain; DNA polymerase delta catalytic subunit; DNA polymerase delta, catalytic subunit; DNA polymerase subunit delta p125; DNA-directed DNA polymerase delta 1; MDPL; POL D; POLD; POLD1; polymerase (DNA directed), delta 1, catalytic subunit; polymerase (DNA directed), delta 1, catalytic subunit 125kDa; polymerase (DNA) delta 1, catalytic subunit; Polymerase D | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 18971, 5424, 59294 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction