Learn More
Invitrogen™ POLD1 Recombinant Rabbit Monoclonal Antibody (24GB4155)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557177
 View more versions of this product
Product Code. 30275417
  • 500.00 € / 100µL
Description
POLD1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes the 125-kDa catalytic subunit of DNA polymerase delta. DNA polymerase delta possesses both polymerase and 3' to 5' exonuclease activity and plays a critical role in DNA replication and repair. Alternatively spliced transcript variants have been observed for this gene, and a pseudogene of this gene is located on the long arm of chromosome 6.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
POLD1
Recombinant Monoclonal
1.5 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O54747, P28340, P52431
POLD1
A synthesized peptide derived from human POLD1 (aa 1000-1065).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB4155
Unconjugated
POLD1
125kDa; CDC2; CDC2 homolog; CRCS10; DAN polymerase delta 1, catalytic subunit; DNA polymerase delta 1, catalytic domain; DNA polymerase delta catalytic subunit; DNA polymerase delta, catalytic subunit; DNA polymerase subunit delta p125; DNA-directed DNA polymerase delta 1; MDPL; POL D; POLD; POLD1; polymerase (DNA directed), delta 1, catalytic subunit; polymerase (DNA directed), delta 1, catalytic subunit 125kDa; polymerase (DNA) delta 1, catalytic subunit; Polymerase D
Rabbit
Affinity chromatography
RUO
18971, 5424, 59294
-20°C