missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLD4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94725-0.02ml
This item is not returnable.
View return policy
Description
POLD4 Polyclonal antibody specifically detects POLD4 in Human samples. It is validated for Western Blot
Specifications
| POLD4 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DNA polymerase delta smallest subunit p12, DNA polymerase delta subunit p12, p12, POLDSDNA polymerase delta subunit 4, polymerase (DNA-directed), delta 4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-107 of human POLD4 (NP_066996.3). MGRKRLITDSYPVVKRREGPAGHSKGELAPELGEEPQPRDEEEAELELLRQFDLAWQYGPCTGITRLQRWCRAKQMGLEPPPEVWQVLKTHPGDPRFQCSLWHLYPL | |
| 0.02 mL | |
| Cell Cycle and Replication | |
| 57804 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction