missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLDIP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
415.00 € - 572.00 €
Specifications
| Antigen | POLDIP2 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18240374
|
Novus Biologicals
NBP2-58909 |
100 μL |
572.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18662127
|
Novus Biologicals
NBP2-58909-25ul |
25 μL |
415.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
POLDIP2 Polyclonal specifically detects POLDIP2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| POLDIP2 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 26073 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:GKVLETVGVFEVPKQNGKYETGQLFLHSIFGYRGVVLFPWQARLYDRDVASAAPEKAENPAGHGSKEVKGKTHTYYQVLIDARDCPHISQRSQTEAVTFLANHDDSRALYAIPGLDYVSHEDILPYTSTDQVPIQ | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| DKFZP586F1524, p38, PDIP38DKFZp586F1524,38 kDa DNA polymerase delta interaction protein, POLD4, polymerase (DNA-directed), delta interacting protein 2, polymerase delta interacting protein 38, polymerase delta-interacting protein 2 | |
| POLDIP2 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title