Learn More
Invitrogen™ RXRG Recombinant Rabbit Monoclonal Antibody (HL3185)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA556859
Product Code. 30275836
  • 500.00 € / 100µL
Description
RXRG Recombinant Monoclonal Antibody for Western Blot
Retinoic Acid (RA; active metabolite of vitamin A) plays a prominent role in regulating the transition of proliferating precursor cells (such as carcinoma cells and neuronal precursors) to postmitotic differentiated cells (Joshi et al., 2005). The Retinoid X Receptors (RXRs) family (RXR alpha, beta, and gamma), preferentially bind 9-cis -RA and regulate gene transcription by forming heterodimers with a second family of RA receptors. RAs have been suggested to potentially play a therapeutic role in cervical cancer (Abu et al., 2005). RAs are known to play key roles in neuronal development and an increasing body of evidence indicates that retinoid signaling may regulate synaptic plasticity and associated learning and memory behaviors (Lane and Bailey, 2005).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RXRG
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
P28705, Q5BJR8
RXRG
Synthetic peptide of human Retinoid X Receptor gamma.
100 μL
Primary
Mouse, Rat
Antibody
IgG
Western Blot
HL3185
Unconjugated
RXRG
Nr2b3; Nuclear receptor subfamily 2 group B member 3; retinoic acid receptor RXR-gamma; retinoic X receptor gamma-1; retinoid X receptor; retinoid X receptor gamma; retinoid X receptor, gamma; RXR gamma; RXR gamma-1; RXRC; RXRG
Rabbit
Protein A
RUO
20183, 83574
-20°C, Avoid Freeze/Thaw Cycles