Learn More
POLR2F Mouse, anti-Human, Clone: 2D4D12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68511-1-IG-20UL
This item is not returnable.
View return policy
Description
The POLR2F antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human POLR2F. This antibody recognizes human, mouse antigen. The POLR2F antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| POLR2F | |
| 2D4D12 | |
| HRBP14.4, POLR2F, POLRF, RPABC14.4, RPABC2, RPB14.4, RPB6, RPB6 homolog, RPC15 | |
| 20 μL | |
| 5435 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG2a |