missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ POLR3D Monoclonal Antibody (4E6)
GREENER_CHOICE

Product Code. 15518732
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15518732 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15518732 Supplier Invitrogen™ Supplier No. MA518491

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Peptide Sequence: GQVVLIKQEK DREAKLAENA CTLADLTEGQ VGKLLIRKSG RVQLLLGKVT LDVTMGTACS FLQELVSVGL GDSRTGEMTV LGHVKHKLVC SPDFESLLDH.

This gene encodes a protein which interacts with the N-terminal region of BRCA1. In addition to its ability to bind BRCA1 in vivo and in vitro, it shares homology with the 2 most conserved regions of BRCA1: the N-terminal RING motif and the C-terminal BRCT domain. The RING motif is a cysteine-rich sequence found in a variety of proteins that regulate cell growth, including the products of tumor suppressor genes and dominant protooncogenes. This protein also contains 3 tandem ankyrin repeats. The BARD1/BRCA1 interaction is disrupted by tumorigenic amino acid substitutions in BRCA1, implying that the formation of a stable complex between these proteins may be an essential aspect of BRCA1 tumor suppression. This protein may be the target of oncogenic mutations in breast or ovarian cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLR3D
Applications ELISA
Classification Monoclonal
Clone 4E6
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Polr3d
Gene Accession No. P05423
Gene Alias 2810426M17Rik; 44kDa; AI326118; AW489084; BN51; BN51 (BHK21) temperature sensitivity complementing; Bn51t; DNA-directed RNA polymerase III 47 kDa polypeptide; DNA-directed RNA polymerase III subunit C4; DNA-directed RNA polymerase III subunit D; DNA-directed RNA polymerase III subunit RPC4; HOX1C; Polr3d; polymerase (RNA) III (DNA directed) polypeptide D; polymerase (RNA) III (DNA directed) polypeptide D, 44kDa; polymerase (RNA) III subunit D; Protein BN51; RNA polymerase III 47 kDa subunit; RNA polymerase III subunit C4; RNA polymerase III subunit D; RPC4; RPC53; RPC53 homolog; temperature sensitive complementation, cell cycle specific, tsBN51; TSBN51
Gene Symbols Polr3d
Host Species Mouse
Immunogen POLR3D (-, 296 a.a. ~ 395 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 661
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.