missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POLR3E Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559585
This item is not returnable.
View return policy
Description
Immunogen sequence: FVMWKFTQSR WVVRKEVATV TKLCAEDVKD FLEHMAVVRI NKGWEFILPY DGEFIKKHPD VVQRQHMLWT GIQAKLEKVY NLVKETMPKK PDAQ Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 93%.
DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. Specific peripheric component of RNA polymerase III which synthesizes small RNAs, such as 5S rRNA and tRNAs. Essential for efficient transcription from both the type 2 VAI and type 3 U6 RNA polymerase III promoters. Plays a key role in sensing and limiting infection by intracellular bacteria and DNA viruses. Acts as nuclear and cytosolic DNA sensor involved in innate immune response. Can sense non-self dsDNA that serves as template for transcription into dsRNA. The non-self RNA polymerase III transcripts, such as Epstein-Barr virus-encoded RNAs (EBERs) induce type I interferon and NF- Kappa-B through the RIG-I pathway.
Specifications
| POLR3E | |
| Polyclonal | |
| Unconjugated | |
| POLR3E | |
| DNA-directed RNA polymerase III 80 kDa polypeptide; DNA-directed RNA polymerase III subunit RPC5; KIAA1452; POLR3E; polymerase (RNA) III (DNA directed) polypeptide E; polymerase (RNA) III (DNA directed) polypeptide E (80kD); polymerase (RNA) III subunit E; RGD1308086; RNA polymerase III 80 kDa subunit RPC5; RNA polymerase III polypeptide E; RNA polymerase III subunit 5; RNA polymerase III subunit C5; RNA polymerase III subunit E; RPC5; Sex-lethal interactor homolog; Sin; Sxl interactor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55718 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NVU0 | |
| POLR3E | |
| Recombinant protein corresponding to Human POLR3E. Recombinant protein control fragment (Product #RP-98554). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction