missing translation for 'onlineSavingsMsg'
Learn More

Poly(2-acrylamido-2-methyl-1-propanesulfonic acid), 10 wt% aq.sol.; ca. MW 800,000

Product Code. 10163770 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10163770 100 g Glass bottle 100g
10062370 25 g Glass bottle 25g
10579623 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10163770 Supplier Thermo Scientific Acros Supplier No. 184961000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 27119-07-9 | (C7H13NO4S)n | 207.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27119-07-9 , 7732-18-5
Molecular Formula (C7H13NO4S)n
Molecular Weight (g/mol) 207.24
MDL Number MFCD00084369
InChI Key XHZPRMZZQOIPDS-UHFFFAOYSA-N
Synonym 2-acrylamido-2-methylpropanesulfonic acid, 2-acrylamido-2-methyl-1-propanesulfonic acid, 2-acrylamide-2-methylpropanesulfonic acid, 1-propanesulfonic acid, 2-methyl-2-1-oxo-2-propenyl amino, polyacrylamidomethylpropane sulfonic acid, unii-490hqe5ki5, 2-acrylamido-2-methylpropanesulfonate, 2-acrylamido-2-methylpropanesulphonic acid, 2-acrylamido-2-methylpropane sulfonic acid, 2-acrylamido-2-methylpropane-1-sulfonic acid
PubChem CID 65360
IUPAC Name 2-methyl-2-(prop-2-enamido)propane-1-sulfonic acid
SMILES CC(C)(CS(O)(=O)=O)NC(=O)C(-*)C-*

Specifications

Physical Form Viscous Liquid
Linear Formula {-CH2CH[CONHC(CH3)2CH2SO3H]-}n
Quantity 100 g
Percent Purity 8.0 to 12.0% (solids)
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Poly(2-acrylamido-2-methyl-1-propanesulfonic acid), 10 wt% aqueous solution
compliance-icons
Product Identifier
  • Poly(2-acrylamido-2-methyl-1-propanesulfonic acid), 10 wt% solution in water
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.