missing translation for 'onlineSavingsMsg'
Learn More

Polyethyleneimine, M.W. 60,000, 50% w/w aq. soln.

Product Code. 15490177
Change view
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15490177 250 g 250g
15480177 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15490177 Supplier Thermo Scientific Alfa Aesar Supplier No. J61270.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A protein precipitant used to purify proteins | CAS: 9002-98-6 | ((C2H5N)zC2H4N)y(C2H5N)x | 43.07 g/mol

Polyethyleneimine is used for protein and nucleic acid precipitation, transfection assays, and to pretreat filters to increase the binding of proteins. Polyethyleneimine can also complex heavy metal ions. An effective permeabilizer of gram negative bacteria. Crosslinked polyethyleneimine is used as an enzyme carrier. Polyethyleneimine cellulose in small columns will separate cyclic nucleotides from related compounds at the same electrolyte concentration that is used for thin layer chromatography on Polyethyleneimine cellulose-coated plates. It is a water soluble polymer used as a membrane lubricant, useful in filtering frozen cell membranes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-98-6
Molecular Formula ((C2H5N)zC2H4N)y(C2H5N)x
Molecular Weight (g/mol) 43.07
MDL Number MFCD00084427
InChI Key NOWKCMXCCJGMRR-UHFFFAOYSA-N
Synonym ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin
PubChem CID 9033
ChEBI CHEBI:30969
IUPAC Name aziridine
SMILES *-CCNCCN(-*)CCN-*

Specifications

Color Colorless to Yellow
Concentration 50% w/w aq. soln.
Density 1.07
Solubility Information It is soluble in water.
Formula Weight 60 kd
Odor Odorless
Quantity 250 g
Physical Form Liquid
Chemical Name or Material Polyethyleneimine
compliance-icons
Product Identifier
  • Polyethyleneimine, M.W. 60,000, 50% w/w aqueous solution
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Polyethyleneimine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.