Learn More
Melford Polymixin B Sulfate
Descripción
Polymyxin B Sulfate is an antibiotic that belongs to the polymyxin class of antibiotics and is primarily effective against Gram-negative bacteria.
Polymyxin B is derived from Bacillus polymyxa and acts by disrupting the integrity of the bacterial cell membrane.
In the field of microbiology and cell culture, Polymyxin B Sulfate is commonly used to prevent bacterial contamination. It is particularly effective against Gram-negative bacteria, including multidrug-resistant strains such as Pseudomonas aeruginosa. Researchers add Polymyxin B to growth media or solutions to inhibit the growth of these bacteria and maintain the purity of cultures.
Overall, Polymyxin B Sulfate is a valuable antibiotic in the life science marketplace, primarily used in microbiology, cell culture, and molecular biology research. Its effectiveness against Gram-negative bacteria and multidrug-resistant strains makes it a crucial tool for maintaining sterile cultures and studying bacterial pathogens. However, careful consideration of its potential toxicity is necessary when using it in cell culture applications.
Especificaciones
Especificaciones
| Chemical Name or Material | Polymixin B Sulfate |
| CAS | 1405-20-5 |
| Quantity | 1 g |
| Molecular Formula | C55H96N16O13-2H2SO4 |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.