missing translation for 'onlineSavingsMsg'
Learn More

Polymixin B sulfate, Cell Culture Reagent

Product Code. 15434289
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15434289 1 g 1g
15444289 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15434289 Supplier Thermo Scientific Alfa Aesar Supplier No. J61763.03
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Polymixin B sulfate, CAS # 1405-20-5, is a short polypeptide with antibacterial activity against gram-negative bacteria. | CAS: 1405-20-5 | C48H84N16O17S | 1189.36 g/mol

In cell culture, polymyxin B sulfate is used as an antibacterial agent to prevent contamination. Polymyxin B sulfate is recommended for use in cell culture applications at 50 mg/L. It is used as a lipid A binding agent that induces pore formation in cell membranes and as immobilized (substrate bound) agent for removal of endotoxins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Polymixin B sulfate is a mixture of B1 and B2 polypeptides obtained from different strains of Bacillus polymyxa, with antibacterial activity against gram-negative bacteria. It does not exhibit activity against fungi and gram-positive organisms
  • This compound inhibits Ca2+ dependent PKC (protein kinase C)
  • It can bind lipopolysaccharide (LPS) of the outer membrane of gram-negative bacteria and disrupts the cytoplasmic membrane by inducing large pores to allow nucleotide leakage in bacterial walls. This disrupts the permeability of the cytoplasmic membrane

Application

  • Polymixin B sulfate targets BAC-1.2F5 murine macrophages as an LPS (lipopolysaccharide) inhibitor
  • In in vitro research, this compound is used to prevent bacterial contamination of cell cultures
  • In microbiology research, it is used as a positive and negative control during microbicidal concentration (MCC) assay
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1405-20-5
Molecular Formula C48H84N16O17S
Molecular Weight (g/mol) 1189.36
MDL Number MFCD00131991
InChI Key HNDFYNOVSOOGDU-UHFFFAOYNA-N
PubChem CID 133109994
SMILES OS(O)(=O)=O.CC(C)CC1NC(=O)C(CC2=CC=CC=C2)NC(=O)C(CCN)NC(=O)C(CCNC(=O)C(CCO)NC(=O)C(CCN)NC(=O)C(CCN)NC1=O)NC(=O)C(CCN)NC(=O)C(CCO)NC(=O)C(CCN)NC=O

Specifications

Type Polymixin B Sulfate
Physical Form Powder
Packaging Glass Bottle
Quantity 1 g
Solubility Information Soluble in water
IUPAC Name N-[(2S)-4-amino-1-[[(2S,3R)-1-[[(2S)-4-amino-1-oxo-1-[[(3S,6S,18R)-6,9,18-tris(2-aminoethyl)-15-benzyl-3-[(1R)-1-hydroxyethyl]-12-(2-methylpropyl)-2,5,8,11,14,17,20-heptaoxo-1,4,7,10,13,16,19-heptazacyclotricos-21-yl]amino]butan-2-yl]amino]-3-hydroxy-1-ox
Formula Weight 1385.63
Chemical Name or Material Polymixin B sulfate
compliance-icons
Product Identifier
  • Polymixin B sulfate, Cell Culture Reagent
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Polymyxin B sulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.