missing translation for 'onlineSavingsMsg'
Learn More

Polymyxin B Sulfate MP Biomedicals

Product Code. 11467660 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 mU
25 mU
5 mU
50 mU
Unit Size:
10 micro units
25 micro units
5 micro units
50 micro units
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11467660 5 mU 5 micro units
11477660 10 mU 10 micro units
11487660 25 mU 25 micro units
11497660 50 mU 50 micro units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11467660 Supplier MP Biomedicals Supplier No. 0210056505
  • ONLINE EXCLUSIVE PRICE
    192.00 €
    181.65 € / 5 micro units
    Save 10.35 €
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 1405-20-5
Molecular Formula C48H84N16O17S
Molecular Weight (g/mol) 1189.36
MDL Number MFCD00131991
InChI Key HNDFYNOVSOOGDU-UHFFFAOYNA-N
Synonym Aerosporin
PubChem CID 133109994
IUPAC Name N-[(2S)-4-amino-1-[[(2S,3R)-1-[[(2S)-4-amino-1-oxo-1-[[(3S,6S,18R)-6,9,18-tris(2-aminoethyl)-15-benzyl-3-[(1R)-1-hydroxyethyl]-12-(2-methylpropyl)-2,5,8,11,14,17,20-heptaoxo-1,4,7,10,13,16,19-heptazacyclotricos-21-yl]amino]butan-2-yl]amino]-3-hydroxy-1-ox
SMILES OS(O)(=O)=O.CC(C)CC1NC(=O)C(CC2=CC=CC=C2)NC(=O)C(CCN)NC(=O)C(CCNC(=O)C(CCO)NC(=O)C(CCN)NC(=O)C(CCN)NC1=O)NC(=O)C(CCN)NC(=O)C(CCO)NC(=O)C(CCN)NC=O

Specifications

Type Polymixin B Sulfate
Physical Form Powder
Color White
Quantity 5 mU
Formula Weight 1385.61
Chemical Name or Material Polymyxin B Sulfate, Aerosporin Activity: >6, 000 u/mg
compliance-icons
Product Identifier
  • Polymyxin B Sulfate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: polymyxin B sulphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.