missing translation for 'onlineSavingsMsg'
Learn More

Polymyxin B sulfate, MP Biomedicals™

Product Code. 11457660 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 mU
Unit Size:
1 micro unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11457660 1 mU 1 micro unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11457660 Supplier MP Biomedicals Supplier No. 0210056501
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Antibiotic used to treat infections caused by Gram-negative bacteria by disrupting the cell membrane.

Specifications

Type Polymixin B Sulfate
Physical Form Powder
CAS 1405-20-5
Color White
Quantity 1 mU
Molecular Formula C48H84N16O17S
MDL Number MFCD00131991
Synonym Aerosporin
InChI Key HNDFYNOVSOOGDU-UHFFFAOYNA-N
SMILES OS(O)(=O)=O.CC(C)CC1NC(=O)C(CC2=CC=CC=C2)NC(=O)C(CCN)NC(=O)C(CCNC(=O)C(CCO)NC(=O)C(CCN)NC(=O)C(CCN)NC1=O)NC(=O)C(CCN)NC(=O)C(CCO)NC(=O)C(CCN)NC=O
IUPAC Name N-[(2S)-4-amino-1-[[(2S,3R)-1-[[(2S)-4-amino-1-oxo-1-[[(3S,6S,18R)-6,9,18-tris(2-aminoethyl)-15-benzyl-3-[(1R)-1-hydroxyethyl]-12-(2-methylpropyl)-2,5,8,11,14,17,20-heptaoxo-1,4,7,10,13,16,19-heptazacyclotricos-21-yl]amino]butan-2-yl]amino]-3-hydroxy-1-ox
Molecular Weight (g/mol) 1189.36
PubChem CID 133109994
Formula Weight 1385.63
Grade Not Available
Chemical Name or Material Polymyxin B sulfate
Show More Show Less
compliance-icons
Product Identifier
  • Polymyxin B Sulfate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.