missing translation for 'onlineSavingsMsg'
Learn More

Polyphenylmethylsiloxane, MW 2500-2700

Product Code. 11475340
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475340 250 g 250g
11465340 50 g 50g
11485340 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11475340 Supplier Thermo Scientific Alfa Aesar Supplier No. 044569.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 9005-12-3 | C7H8OSi | 136.23 g/mol

A biochemical for proteomics research. A novel coating for solid-phase microextraction. A polyphenylmethylsiloxane for use in skincare, suncare, color cosmetic, hair care, and antiperspirant / deodorant applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-12-3
Molecular Weight (g/mol) 136.23
MDL Number MFCD00146731

Specifications

Physical Form Liquid
Molecular Formula C7H8OSi
Synonym methylphenylsilanone, polyphenylmethylsiloxane, methylphenyl oxo silane, silane, methyloxophenyl, methyl phenyl silanone
Solubility Information Insoluble in water.
Quantity 250 g
InChI Key IXKNJYSAWNJQQY-UHFFFAOYSA-N
SMILES C[Si](=O)C1=CC=CC=C1
IUPAC Name methyl-oxo-phenylsilane
PubChem CID 3032548
Flash Point 300°C (572°F)
Chemical Name or Material Polyphenylmethylsiloxane, M.W. 2,500 to 2,700
Show More Show Less

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.