missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ twin.tec™ PCR Plates

Product Code. 10690744 Shop All Eppendorf Products
Change view
Click to view available options
Color:
Blue
Clear
Red
Skirt Style:
Semi
Skirted
Unit Size:
Pack of 25
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Skirt Style Unit Size
10690744 Clear Semi Pack of 25
10767294 Blue Semi Pack of 25
10355672 Blue Skirted Pack of 25
10120092 Clear Skirted Pack of 25
10100803 Red Skirted Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10690744 Supplier Eppendorf™ Supplier No. 0030128575
  • ONLINE EXCLUSIVE PRICE
    166.00 €
    154.00 € / Pack of 25
    Save 12.00 €
    7% Off
In Stock
Add to basket
Edge
Add to basket
Bulk Quote Request

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use these plates for automation and high throughput processes as well as quantitative real-time PCR. Eppendorf™ twin.tec™ PCR Plates combine extremely thin walled polypropylene wells for optimal heat transfer and extremely robust polycarbonate frames for rigidity and torque-resistance.

  • Raised well rims for effective sealing, also reduces risk of cross-contamination
  • Exceptionally solid and torque-resistant polycarbonate frame
  • Improved well-to-well tolerance
  • Cut-away corner and alphanumeric grid-referencing
  • Certified free of any detectable human DNA, DNase, RNase and PCR inhibitors
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Packaging 25 Pk.
Binding Type Minimal
Product Line Eppendorf
Color Clear
Disposable Single-use
Product Type PCR Microplate
For Use With (Equipment) Real time PCR
Skirt Style Semi
Material Polycarbonate
Sterility Sterile
Total Volume (Metric) Well 150 μL
Well Shape Conical Bottom (V-well)
Dnase Free Yes
For Use With (Application) Ideal for quantitative real time PCR and fit most thermal cyclers
Quantity 25/Pk.
RNase Free Yes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.