missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Polystyrene Microplates
Designed for in vitro diagnostic assays
Brand: Corning™ 2510
Code : 67
Additional Details : Weight : 0.02239kg
Description
The Stripwell™ low volume microplates allow test kit manufacturers to reduce overall costs, with a reduction in the amount of reagents and antibody necessary to perform assays.- Designed with one-half the well area of a 'regular' 8 well strip, providing a much greater surface to volume ratio
- Recommended working volume of 50μL to 125μL
- Designed for in vitro diagnostic assays
- Untreated surface is hydrophobic in nature
- Color coded strips
- Bar coding available on request
Specifications
Flat | |
Without Lid | |
Clear | |
96 | |
Non-sterile | |
360 μL | |
1 Pack | |
Clear |
Strip Plate | |
8 x 12 | |
Polystyrene | |
75 to 200 μL | |
Sulfhydryl | |
50 | |
50/Cs. |