Learn More
Greiner Bio-One Polystyrene Standard Lids for Microplates
Product Code. 10761211
Change view
Click to view available options
Plate Compatibility:
384-well Standard Microplate
96-well Microplate, Non TC-treated
High-format Low-profile Microplates, 384-well LoBase Small Volume or 1536-well Microplate
Sterility:
Non-sterile
Sterile
Format:
High profile
Low profile
Ultra low profile
Unit Size:
Pack of 100
Pack of 200
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plate Compatibility Sterility Format Unit Size
10761211 96-well Microplate, Non TC-treated Sterile High profile Pack of 100
10389031 384-well Standard Microplate Non-sterile Low profile Pack of 200
10421611 96-well Microplate, Non TC-treated Non-sterile High profile Pack of 100
10414625 High-format Low-profile Microplates, 384-well LoBase Small Volume or 1536-well Microplate Sterile Ultra low profile Pack of 100
10292811 384-well Standard Microplate Sterile Low profile Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10761211 Supplier Greiner Bio-One Supplier No. 656161
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Provides protection against contamination and evaporation during sample storage and cell cultivation. Greiner BioOne™ Polystyrene Standard Lids are available in a wide variety of formats to meet laboratory needs.
  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic
  • High lid profile; height: 9 mm
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Material Polystyrene
Sterility Sterile
Includes Chimney-well style 96 well plates
Condensation Rings No
No. per Pack 100
Format High profile
Product Type Plate Lid
Plate Compatibility 96-well Microplate, Non TC-treated