missing translation for 'onlineSavingsMsg'
Learn More

Polysucrose 400, MP Biomedicals™

Product Code. 11463692 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11463692 100 g 100g
11443692 10 g 10g
11453692 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11463692 Supplier MP Biomedicals Supplier No. 0215632880
 Restricted Item
Estimated Shipment: 11-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 26873-85-8
Molecular Formula C15H27ClO12
Molecular Weight (g/mol) 434.819
MDL Number MFCD00081599
InChI Key IACFXVUNKCXYJM-AKSHDPDZSA-N
Synonym ficol 400
PubChem CID 88330506
IUPAC Name 2-(chloromethyl)oxirane;(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C1C(O1)CCl.C(C1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)CO)O)O)O)O

Specifications

Color White
Physical Form Powder
Quantity 100 g
Formula Weight ≈400,000
Chemical Name or Material Polysucrose 400
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.