missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PON1 Antibody (SR1662), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22058-50ul
This item is not returnable.
View return policy
Description
PON1 Monoclonal antibody specifically detects PON1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation
Specifications
| PON1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| SR1662 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 | |
| A-esterase 1, Aromatic esterase 1, arylesterase B-type, EC 3.1.1.2, EC 3.1.1.81, EC 3.1.8.1, ESAesterase A, K-45, MVCD5, paraoxonase 1, PON 1, PONparaoxonase B-type, serum aryldiakylphosphatase, Serum aryldialkylphosphatase 1, serum paraoxonase/arylesterase 1 | |
| A synthesized peptide derived from human PON1 (Uniprot #: P27169) | |
| 50 μg | |
| Alzheimers Research, Cancer, Lipid and Metabolism, Neurodegeneration, Neuroscience | |
| 5444 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction