missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PON2 Recombinant Rabbit Monoclonal Antibody (24GB1035)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557536
This item is not returnable.
View return policy
Description
PON2 Recombinant Monoclonal Antibody for Western Blot, Flow
This gene encodes a member of the paraoxonase gene family, which includes three known members located adjacent to each other on the long arm of chromosome 7. The encoded protein is ubiquitously expressed in human tissues, membrane-bound, and may act as a cellular antioxidant, protecting cells from oxidative stress. Hydrolytic activity against acylhomoserine lactones, important bacterial quorum-sensing mediators, suggests the encoded protein may also play a role in defense responses to pathogenic bacteria. Mutations in this gene may be associated with vascular disease and a number of quantitative phenotypes related to diabetes. Alternatively spliced transcript variants encoding different isoforms have been described.
Specifications
| PON2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q15165 | |
| PON2 | |
| A synthesized peptide derived from human PON2 (aa 200-354). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1035 | |
| Unconjugated | |
| PON2 | |
| 6330405I24Rik; A esterase 2; A-esterase 2; AI481612; aromatic esterase 2; arylesterase 2; paraoxonase 2; paraoxonase nirs; PON 2; PON2; Serum aryldialkylphosphatase 2; serum paraoxonase/arylesterase 2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 5445 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction