missing translation for 'onlineSavingsMsg'
Learn More

Ponceau 4RC (E124) 100 μg/mL in Acetonitrile:Water, Dr. Ehrenstorfer

Product Code. 30324081
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30324081 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30324081 Supplier Dr. Ehrenstorfer Supplier No. DREA16284000WL100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ponceau 4RC (E124)
CAS 2611-82-7
Analyte or Component Names Ponceau 4RC (E124)
Shipping Condition Room Temperature
Molecular Formula C20 H11 N2 O10 S3 . 3 Na
Concentration 100 μg/mL
Certifications/Compliance ISO/IEC 17025
Synonym GW 306684A
InChI Formula InChI=1S/C20H14N2O10S3.3Na/c23-16-7-5-11-9-12(33(24,25)26)10-18(35(30,31)32)19(11)20(16)22-21-15-6-8-17(34(27,28)29)14-4-2-1-3-13(14)15;;;/h1-10,23H,(H,24,25,26)(H,27,28,29)(H,30,31,32);;;/q;3*+1/p-3
SMILES [Na+].[Na+].[Na+].Oc1ccc2cc(cc(c2c1N=Nc3ccc(c4ccccc34)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-]
IUPAC Name trisodium;7-hydroxy-8-[(E)-(4-sulfonatonaphthalen-1-yl)diazenyl]naphthalene-1,3-disulfonate
Solvent or Matrix Acetonitrile/Water
Molecular Weight (g/mol) 604.47
Quantity 1 mL
Formula Weight 603.9269 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.